Assay Details
Gene Symbol: LOC5564121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855336.1 | XP_021711028.1 | 1 - 2 | 497 | 78 | |
XM_021855337.1 | XP_021711029.1 | 1 - 2 | 492 | 78 | ||
XM_021855338.1 | XP_021711030.1 | 1 - 2 | 492 | 78 |
