Assay Details
Gene Symbol: LOC5572780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654128.2 | XP_001654178.2 | 1 - 2 | 187 | 111 | |
XM_021852461.1 | XP_021708153.1 | 1 - 2 | 187 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001654128.2, XM_021852461.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
