Assay Details
Gene Symbol: LOC5568456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652239.2 | XP_001652289.1 | 2 - 3 | 912 | 82 | |
XM_021850951.1 | XP_021706643.1 | 2 - 3 | 902 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
