Assay Details
Gene Symbol: LOC5575737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840026.1 | XP_021695718.1 | 4 - 5 | 2026 | 58 | |
XM_021840027.1 | XP_021695719.1 | 5 - 6 | 1933 | 58 | ||
XM_021840029.1 | XP_021695721.1 | 5 - 6 | 1561 | 58 | ||
XM_021840030.1 | XP_021695722.1 | 4 - 5 | 2026 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
