Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 13 - 14 | 4117 | 96 | |
XM_021845538.1 | XP_021701230.1 | 13 - 14 | 4072 | 96 | ||
XM_021845539.1 | XP_021701231.1 | 12 - 13 | 3846 | 96 | ||
XM_021845540.1 | XP_021701232.1 | 12 - 13 | 2917 | 96 | ||
XM_021845541.1 | XP_021701233.1 | 12 - 13 | 2872 | 96 | ||
XM_021845542.1 | XP_021701234.1 | 13 - 14 | 4117 | 96 | ||
XM_021845543.1 | XP_021701235.1 | 11 - 12 | 2581 | 96 | ||
XM_021845544.1 | XP_021701236.1 | 11 - 12 | 2536 | 96 | ||
XM_021845545.1 | XP_021701237.1 | 2 - 3 | 270 | 96 |
