Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 17 - 18 | 6028 | 70 | |
XM_021845538.1 | XP_021701230.1 | 17 - 18 | 5983 | 70 | ||
XM_021845539.1 | XP_021701231.1 | 16 - 17 | 5757 | 70 | ||
XM_021845540.1 | XP_021701232.1 | 16 - 17 | 4828 | 70 | ||
XM_021845541.1 | XP_021701233.1 | 16 - 17 | 4783 | 70 | ||
XM_021845542.1 | XP_021701234.1 | 16 - 17 | 4675 | 70 | ||
XM_021845543.1 | XP_021701235.1 | 15 - 16 | 4492 | 70 | ||
XM_021845544.1 | XP_021701236.1 | 15 - 16 | 4447 | 70 | ||
XM_021845545.1 | XP_021701237.1 | 6 - 7 | 2181 | 70 |

More Information
Set Membership: |
![]() ![]() |
