Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 18 - 19 | 6118 | 66 | |
XM_021845538.1 | XP_021701230.1 | 18 - 19 | 6073 | 66 | ||
XM_021845539.1 | XP_021701231.1 | 17 - 18 | 5847 | 66 | ||
XM_021845540.1 | XP_021701232.1 | 17 - 18 | 4918 | 66 | ||
XM_021845541.1 | XP_021701233.1 | 17 - 18 | 4873 | 66 | ||
XM_021845542.1 | XP_021701234.1 | 17 - 18 | 4765 | 66 | ||
XM_021845543.1 | XP_021701235.1 | 16 - 17 | 4582 | 66 | ||
XM_021845544.1 | XP_021701236.1 | 16 - 17 | 4537 | 66 | ||
XM_021845545.1 | XP_021701237.1 | 7 - 8 | 2271 | 66 |

More Information
Set Membership: |
![]() ![]() |
