Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 21 - 22 | 6598 | 79 | |
XM_021845538.1 | XP_021701230.1 | 21 - 22 | 6553 | 79 | ||
XM_021845539.1 | XP_021701231.1 | 20 - 21 | 6327 | 79 | ||
XM_021845540.1 | XP_021701232.1 | 20 - 21 | 5398 | 79 | ||
XM_021845541.1 | XP_021701233.1 | 20 - 21 | 5353 | 79 | ||
XM_021845542.1 | XP_021701234.1 | 20 - 21 | 5245 | 79 | ||
XM_021845543.1 | XP_021701235.1 | 19 - 20 | 5062 | 79 | ||
XM_021845544.1 | XP_021701236.1 | 19 - 20 | 5017 | 79 | ||
XM_021845545.1 | XP_021701237.1 | 10 - 11 | 2751 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
