Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 1 - 2 | 670 | 63 | |
XM_021845538.1 | XP_021701230.1 | 1 - 2 | 670 | 63 | ||
XM_021845540.1 | XP_021701232.1 | 1 - 2 | 670 | 63 | ||
XM_021845541.1 | XP_021701233.1 | 1 - 2 | 670 | 63 | ||
XM_021845542.1 | XP_021701234.1 | 1 - 2 | 670 | 63 | ||
XM_021845543.1 | XP_021701235.1 | 1 - 2 | 670 | 63 | ||
XM_021845544.1 | XP_021701236.1 | 1 - 2 | 670 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845536.1, XM_021845538.1, XM_021845540.1, XM_021845541.1, XM_021845542.1, XM_021845543.1, XM_021845544.1 transcripts.
Set Membership: |
![]() ![]() |
