Assay Details
Gene Symbol: LOC5567456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845536.1 | XP_021701228.1 | 2 - 3 | 1043 | 71 | |
XM_021845538.1 | XP_021701230.1 | 2 - 3 | 1043 | 71 | ||
XM_021845540.1 | XP_021701232.1 | 2 - 3 | 1043 | 71 | ||
XM_021845541.1 | XP_021701233.1 | 2 - 3 | 1043 | 71 | ||
XM_021845542.1 | XP_021701234.1 | 2 - 3 | 1043 | 71 | ||
XM_021845543.1 | XP_021701235.1 | 2 - 3 | 1043 | 71 | ||
XM_021845544.1 | XP_021701236.1 | 2 - 3 | 1043 | 71 |
