Assay Details
Gene Symbol: LOC5567089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841760.1 | XP_021697452.1 | 5 - 6 | 2153 | 74 | |
XM_021841761.1 | XP_021697453.1 | 5 - 6 | 2152 | 74 | ||
XM_021841762.1 | XP_021697454.1 | 5 - 6 | 2026 | 74 | ||
XM_021841763.1 | XP_021697455.1 | 6 - 7 | 2260 | 74 | ||
XM_021841764.1 | XP_021697456.1 | 6 - 7 | 2240 | 74 | ||
XM_021841765.1 | XP_021697457.1 | 5 - 6 | 2059 | 74 | ||
XM_021841766.1 | XP_021697458.1 | 5 - 6 | 2033 | 74 | ||
XM_021841767.1 | XP_021697459.1 | 5 - 6 | 1941 | 74 | ||
XM_021841768.1 | XP_021697460.1 | 5 - 6 | 1939 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
