Assay Details
Gene Symbol: LOC5567089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841760.1 | XP_021697452.1 | 3 - 4 | 1069 | 62 | |
XM_021841761.1 | XP_021697453.1 | 3 - 4 | 1068 | 62 | ||
XM_021841762.1 | XP_021697454.1 | 3 - 4 | 942 | 62 | ||
XM_021841763.1 | XP_021697455.1 | 4 - 5 | 1176 | 62 | ||
XM_021841764.1 | XP_021697456.1 | 4 - 5 | 1156 | 62 | ||
XM_021841765.1 | XP_021697457.1 | 3 - 4 | 975 | 62 | ||
XM_021841766.1 | XP_021697458.1 | 3 - 4 | 949 | 62 | ||
XM_021841767.1 | XP_021697459.1 | 3 - 4 | 857 | 62 | ||
XM_021841768.1 | XP_021697460.1 | 3 - 4 | 855 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
