Assay Details
Gene Symbol: LOC110678644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851779.1 | XP_021707471.1 | 2 - 3 | 1219 | 66 | |
XM_021851780.1 | XP_021707472.1 | 2 - 3 | 1202 | 66 |

More Information
Set Membership: |
![]() ![]() |
