Assay Details
Gene Symbol: LOC110678644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851779.1 | XP_021707471.1 | 4 - 5 | 1601 | 53 | |
XM_021851781.1 | XP_021707473.1 | 4 - 5 | 1587 | 53 |

More Information
Set Membership: |
![]() ![]() |
