Assay Details
Gene Symbol: LOC110678644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851779.1 | XP_021707471.1 | 5 - 6 | 1682 | 106 | |
XM_021851780.1 | XP_021707472.1 | 5 - 6 | 1647 | 106 | ||
XM_021851781.1 | XP_021707473.1 | 5 - 6 | 1668 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
