Assay Details
Gene Symbol: LOC5565411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841926.1 | XP_021697618.1 | 4 - 5 | 4220 | 57 | |
XM_021841927.1 | XP_021697619.1 | 4 - 5 | 4219 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
