Assay Details
Gene Symbol: LOC5573800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848508.1 | XP_021704200.1 | 15 - 16 | 6916 | 68 | |
XM_021848509.1 | XP_021704201.1 | 15 - 16 | 5111 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
