Assay Details
Gene Symbol: LOC5573800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848508.1 | XP_021704200.1 | 12 - 13 | 6202 | 76 | |
XM_021848509.1 | XP_021704201.1 | 12 - 13 | 4397 | 76 |
