Assay Details
Gene Symbol: LOC5573800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848508.1 | XP_021704200.1 | 11 - 12 | 5925 | 59 | |
XM_021848509.1 | XP_021704201.1 | 11 - 12 | 4120 | 59 |

More Information
Set Membership: |
![]() ![]() |
