Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845509.1 | XP_021701201.1 | 6 - 7 | 2100 | 75 | |
XM_021845510.1 | XP_021701202.1 | 7 - 8 | 1997 | 75 | ||
XM_021845511.1 | XP_021701203.1 | 7 - 8 | 1807 | 75 | ||
XM_021845512.1 | XP_021701204.1 | 7 - 8 | 1902 | 75 | ||
XM_021845513.1 | XP_021701205.1 | 6 - 7 | 2102 | 75 | ||
XM_021845514.1 | XP_021701206.1 | 6 - 7 | 2100 | 75 | ||
XM_021845515.1 | XP_021701207.1 | 6 - 7 | 2100 | 75 | ||
XM_021845516.1 | XP_021701208.1 | 6 - 7 | 2101 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
