Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845515.1 | XP_021701207.1 | 9 - 10 | 4384 | 79 | |
XM_021845516.1 | XP_021701208.1 | 9 - 10 | 4358 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
