Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845509.1 | XP_021701201.1 | 13 - 14 | 5215 | 95 | |
XM_021845510.1 | XP_021701202.1 | 14 - 15 | 5112 | 95 | ||
XM_021845511.1 | XP_021701203.1 | 14 - 15 | 4922 | 95 | ||
XM_021845512.1 | XP_021701204.1 | 14 - 15 | 5017 | 95 | ||
XM_021845513.1 | XP_021701205.1 | 13 - 14 | 5190 | 95 | ||
XM_021845514.1 | XP_021701206.1 | 13 - 14 | 5215 | 95 | ||
XM_021845515.1 | XP_021701207.1 | 14 - 15 | 5149 | 95 | ||
XM_021845516.1 | XP_021701208.1 | 14 - 15 | 5123 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
