Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845514.1 | XP_021701206.1 | 14 - 15 | 5848 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845514.1 | XP_021701206.1 | 14 - 15 | 5848 | 54 |
Set Membership: |
![]() ![]() ![]() ![]() |