Assay Details
Gene Symbol: LOC5571651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845509.1 | XP_021701201.1 | 1 - 2 | 966 | 72 | |
XM_021845510.1 | XP_021701202.1 | 2 - 3 | 863 | 72 | ||
XM_021845511.1 | XP_021701203.1 | 2 - 3 | 673 | 72 | ||
XM_021845512.1 | XP_021701204.1 | 2 - 3 | 768 | 72 | ||
XM_021845513.1 | XP_021701205.1 | 1 - 2 | 968 | 72 | ||
XM_021845514.1 | XP_021701206.1 | 1 - 2 | 966 | 72 | ||
XM_021845515.1 | XP_021701207.1 | 1 - 2 | 966 | 72 | ||
XM_021845516.1 | XP_021701208.1 | 1 - 2 | 967 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845509.1, XM_021845510.1, XM_021845511.1, XM_021845512.1, XM_021845513.1, XM_021845514.1, XM_021845515.1, XM_021845516.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
