Assay Details
Gene Symbol: LOC5575110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661782.2 | XP_001661832.1 | 2 - 3 | 285 | 62 | |
XM_021851003.1 | XP_021706695.1 | 3 - 4 | 986 | 62 | ||
XM_021851004.1 | XP_021706696.1 | 3 - 4 | 986 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001661782.2, XM_021851003.1, XM_021851004.1 transcripts.
Set Membership: |
![]() ![]() |
