Assay Details
Gene Symbol: LOC5575110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661782.2 | XP_001661832.1 | 10 - 11 | 2107 | 76 | |
XM_021851003.1 | XP_021706695.1 | 11 - 12 | 2808 | 76 | ||
XM_021851004.1 | XP_021706696.1 | 11 - 12 | 2712 | 76 |
