Assay Details
Gene Symbol: LOC5575110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661782.2 | XP_001661832.1 | 6 - 7 | 980 | 76 | |
XM_021851003.1 | XP_021706695.1 | 7 - 8 | 1681 | 76 | ||
XM_021851004.1 | XP_021706696.1 | 7 - 8 | 1585 | 76 |
