Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847232.1 | XP_021702924.1 | 6 - 7 | 1256 | 86 | |
XM_021847233.1 | XP_021702925.1 | 8 - 9 | 2614 | 86 | ||
XM_021847234.1 | XP_021702926.1 | 7 - 8 | 1849 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
