Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847232.1 | XP_021702924.1 | 5 - 6 | 1136 | 63 | |
XM_021847233.1 | XP_021702925.1 | 7 - 8 | 2494 | 63 | ||
XM_021847234.1 | XP_021702926.1 | 6 - 7 | 1729 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021847232.1, XM_021847233.1, XM_021847234.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
