Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847233.1 | XP_021702925.1 | 3 - 4 | 1447 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021847233.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
