Assay Details
Gene Symbol: LOC5571858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847232.1 | XP_021702924.1 | 12 - 13 | 2172 | 63 | |
XM_021847233.1 | XP_021702925.1 | 14 - 15 | 3530 | 63 | ||
XM_021847234.1 | XP_021702926.1 | 13 - 14 | 2765 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
