Assay Details
Gene Symbol: LOC5577190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846019.1 | XP_021701711.1 | 3 - 4 | 1012 | 93 | |
XM_021846020.1 | XP_021701712.1 | 3 - 4 | 1012 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
