Assay Details
Gene Symbol: LOC5577190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846019.1 | XP_021701711.1 | 1 - 2 | 145 | 140 | |
XM_021846020.1 | XP_021701712.1 | 1 - 2 | 145 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021846019.1, XM_021846020.1 transcripts.
Set Membership: |
![]() ![]() |
