Assay Details
Gene Symbol: LOC5578886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001664044.2 | XP_001664094.1 | 4 - 5 | 1448 | 79 | |
XM_021839620.1 | XP_021695312.1 | 5 - 6 | 1383 | 79 | ||
XM_021839625.1 | XP_021695317.1 | 5 - 6 | 1224 | 79 | ||
XM_021839629.1 | XP_021695321.1 | 5 - 6 | 1184 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
