Assay Details
Gene Symbol: LOC5576526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662670.2 | XP_001662720.1 | 1 - 2 | 332 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001662670.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
