Assay Details
Gene Symbol: LOC5573509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654598.2 | XP_001654648.1 | 2 - 3 | 1118 | 79 | |
XM_021846555.1 | XP_021702247.1 | 3 - 4 | 1149 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
