Assay Details
Gene Symbol: LOC5573710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843229.1 | XP_021698921.1 | 5 - 6 | 2803 | 100 | |
XM_021843230.1 | XP_021698922.1 | 4 - 5 | 1993 | 100 | ||
XM_021843231.1 | XP_021698923.1 | 6 - 7 | 2600 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
