Assay Details
Gene Symbol: LOC5573710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843229.1 | XP_021698921.1 | 3 - 4 | 1544 | 115 | |
XM_021843230.1 | XP_021698922.1 | 2 - 3 | 734 | 115 | ||
XM_021843231.1 | XP_021698923.1 | 4 - 5 | 1341 | 115 |

More Information
Set Membership: |
![]() ![]() |
