Assay Details
Gene Symbol: LOC5573710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843230.1 | XP_021698922.1 | 1 - 2 | 219 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843230.1 transcripts.
Set Membership: |
![]() ![]() |
