Assay Details
Gene Symbol: LOC23687646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011495202.2 | XP_011493504.1 | 2 - 3 | 475 | 68 | |
XM_021850360.1 | XP_021706052.1 | 2 - 3 | 508 | 68 | ||
XM_021850361.1 | XP_021706053.1 | 2 - 3 | 428 | 68 | ||
XM_021850363.1 | XP_021706055.1 | 2 - 3 | 461 | 68 | ||
XM_021850364.1 | XP_021706056.1 | 2 - 3 | 420 | 68 | ||
XM_021850365.1 | XP_021706057.1 | 2 - 3 | 271 | 68 | ||
XM_021850366.1 | XP_021706058.1 | 2 - 3 | 290 | 68 | ||
XM_021850367.1 | XP_021706059.1 | 2 - 3 | 257 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
