Assay Details
Gene Symbol: LOC23687646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011495202.2 | XP_011493504.1 | 3 - 4 | 1202 | 57 | |
XM_021850360.1 | XP_021706052.1 | 3 - 4 | 1235 | 57 | ||
XM_021850361.1 | XP_021706053.1 | 3 - 4 | 1155 | 57 | ||
XM_021850363.1 | XP_021706055.1 | 3 - 4 | 1188 | 57 | ||
XM_021850364.1 | XP_021706056.1 | 3 - 4 | 1147 | 57 | ||
XM_021850365.1 | XP_021706057.1 | 3 - 4 | 998 | 57 | ||
XM_021850366.1 | XP_021706058.1 | 3 - 4 | 1017 | 57 | ||
XM_021850367.1 | XP_021706059.1 | 3 - 4 | 984 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
