Assay Details
Gene Symbol: LOC23687646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011495202.2 | XP_011493504.1 | 4 - 5 | 2828 | 66 | |
XM_021850360.1 | XP_021706052.1 | 4 - 5 | 2861 | 66 | ||
XM_021850361.1 | XP_021706053.1 | 4 - 5 | 2781 | 66 | ||
XM_021850363.1 | XP_021706055.1 | 4 - 5 | 2814 | 66 | ||
XM_021850364.1 | XP_021706056.1 | 4 - 5 | 2773 | 66 | ||
XM_021850365.1 | XP_021706057.1 | 4 - 5 | 2624 | 66 | ||
XM_021850366.1 | XP_021706058.1 | 4 - 5 | 2643 | 66 | ||
XM_021850367.1 | XP_021706059.1 | 4 - 5 | 2610 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
