Assay Details
Gene Symbol: LOC110674164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838046.1 | XP_021693738.1 | 4 - 5 | 1326 | 80 | |
XM_021838047.1 | XP_021693739.1 | 4 - 5 | 1349 | 80 | ||
XM_021838048.1 | XP_021693740.1 | 3 - 4 | 1216 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
