Assay Details
Gene Symbol: LOC110674164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838046.1 | XP_021693738.1 | 1 - 2 | 261 | 66 | |
XM_021838048.1 | XP_021693740.1 | 1 - 2 | 262 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021838046.1, XM_021838048.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
