Assay Details
Gene Symbol: LOC5570347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653126.2 | XP_001653176.2 | 5 - 6 | 1048 | 92 | |
XM_021843773.1 | XP_021699465.1 | 6 - 7 | 2434 | 92 | ||
XM_021843774.1 | XP_021699466.1 | 7 - 8 | 2340 | 92 | ||
XM_021843775.1 | XP_021699467.1 | 7 - 8 | 2153 | 92 | ||
XM_021843776.1 | XP_021699468.1 | 6 - 7 | 2435 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
