Assay Details
Gene Symbol: LOC5570347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653126.2 | XP_001653176.2 | 7 - 8 | 1309 | 68 | |
XM_021843773.1 | XP_021699465.1 | 8 - 9 | 2695 | 68 | ||
XM_021843774.1 | XP_021699466.1 | 9 - 10 | 2601 | 68 | ||
XM_021843775.1 | XP_021699467.1 | 9 - 10 | 2414 | 68 | ||
XM_021843776.1 | XP_021699468.1 | 8 - 9 | 2696 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
