Assay Details
Gene Symbol: LOC5564087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850753.1 | XP_021706445.1 | 14 - 15 | 3018 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5564087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850753.1 | XP_021706445.1 | 14 - 15 | 3018 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() |