Assay Details
Gene Symbol: LOC5570016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658856.2 | XP_001658906.1 | 3 - 4 | 1203 | 58 | |
XM_021853283.1 | XP_021708975.1 | 3 - 4 | 1192 | 58 | ||
XM_021853284.1 | XP_021708976.1 | 3 - 4 | 1149 | 58 | ||
XM_021853285.1 | XP_021708977.1 | 3 - 4 | 1149 | 58 | ||
XM_021853286.1 | XP_021708978.1 | 3 - 4 | 1124 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
