Assay Details
Gene Symbol: LOC5570016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658856.2 | XP_001658906.1 | 2 - 3 | 348 | 67 | |
XM_021853283.1 | XP_021708975.1 | 2 - 3 | 337 | 67 | ||
XM_021853284.1 | XP_021708976.1 | 2 - 3 | 294 | 67 | ||
XM_021853285.1 | XP_021708977.1 | 2 - 3 | 294 | 67 | ||
XM_021853286.1 | XP_021708978.1 | 2 - 3 | 269 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001658856.2, XM_021853283.1, XM_021853284.1, XM_021853285.1, XM_021853286.1 transcripts.
Set Membership: |
![]() ![]() |
