Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 10 - 11 | 1647 | 61 | |
XM_021845079.1 | XP_021700771.1 | 10 - 11 | 1603 | 61 | ||
XM_021845080.1 | XP_021700772.1 | 9 - 10 | 1279 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
